Learn More
Research Products International Corp Memory Stopwatch Timer
SDP
Catalog No. 501978389
Encompass
Counts up or down for 10 hours in intervals of 1 second. In count down mode unique feature tells you how much time has elapsed after the alarm sounds. Memory allows repeat of just completed cycle. Alarm clock mode. Has a built-in stand, pocket clip, and magnet. A button cell battery is included.
5 Year Warranty
Catalog No. 50-197-8389 Supplier Research Products International Corp Supplier No. 810029
Counts up or down for 10 hours in intervals of 1 second. In count down mode unique feature tells you how much time has elapsed after the alarm sounds. Memory allows repeat of just completed cycle. Alarm clock mode. Has a built-in stand, pocket clip, and magnet. A button cell battery is included.
5 Year Warranty