Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Memory Test Solution 1 For ICP/MS, SPEX CertiPrep™
SDP

Catalog No. CLMEM1 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CL-MEM-1 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CL-MEM-1 Supplier SPEX CertiPrep Supplier No. CLMEM1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al1000μg/mL
Ca1000μg/mL
Fe1000μg/mL
K1000μg/mL
Mg1000μg/mL
Na1000μg/mL
Ag20μg/mL
As20μg/mL
Ba20μg/mL
Be20μg/mL
Cd20μg/mL
Co20μg/mL
Cr20μg/mL
Cu20μg/mL
Mn20μg/mL
Ni20μg/mL
Pb20μg/mL
Se20μg/mL
Tl20μg/mL
V20μg/mL
Zn20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Memory Test 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Solvent or Matrix 5% HNO3
Quantity 125 mL
Grade ICP/ICP-MS
Concentration or Composition (by Analyte or Components) Components in 5% HNO3:
Al    •1000μg/mL
As    •20μg/mL
Ba    •20μg/mL
Be    •20μg/mL
Cd    •20μg/mL
Ca    •1000μg/mL
Co    •20μg/mL
Cr    •20μg/mL
Cu    •20μg/mL
Fe    •1000μg/mL
Pb    •20μg/mL
Mg    •1000μg/mL
Mn    •20μg/mL
Ni    •20μg/mL
K    •1000μg/mL
Se    •20μg/mL
Ag    •20μg/mL
Na    •1000μg/mL
Tl    •20μg/mL
V    •20μg/mL
Zn    •20μg/mL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.