Learn More
MSC MCR Safety General Purpose Gloves, Pigskin Leather, Unlined, X-Large, 1 Pair
Catalog No. 5000684073
Encompass
These general purpose driver-style work gloves are constructed from select B grade grain pigskin leather for reliable durability in professional applications. The naturally porous hide provides superior breathability, and the leather remains soft even after water exposure. Designed for tasks requiring dexterity and precision, these unlined gloves feature a slim profile for an improved hand feel.
  • Constructed from select grade grain pigskin for abrasion resistance
  • Unlined interior provides a low-profile fit and maximum dexterity
  • Naturally porous hide offers superior breathability compared to cowhide
  • Straight thumb design promotes a natural hand position
  • Shirred elastic back keeps the glove securely in place
  • Color-coded hem allows for fast size identification
  • Suitable for general material handling, light assembly, and trade applications
Catalog No. 50-006-84073 Supplier MSC Supplier No. 37563640
These general purpose driver-style work gloves are constructed from select B grade grain pigskin leather for reliable durability in professional applications. The naturally porous hide provides superior breathability, and the leather remains soft even after water exposure. Designed for tasks requiring dexterity and precision, these unlined gloves feature a slim profile for an improved hand feel.
  • Constructed from select grade grain pigskin for abrasion resistance
  • Unlined interior provides a low-profile fit and maximum dexterity
  • Naturally porous hide offers superior breathability compared to cowhide
  • Straight thumb design promotes a natural hand position
  • Shirred elastic back keeps the glove securely in place
  • Color-coded hem allows for fast size identification
  • Suitable for general material handling, light assembly, and trade applications