Learn More
Molecular Dimensions MEMSTART + MEMSYS HT-96
SDP
Catalog No. NC9404328
Encompass
Protein Crystallography. MEMSTART + MEMSYS Screen HT-96 (96 x 1.1mL) For transmembrane proteins using vapour diffusion methods, allows the pH range, precipitants and salts used in membrane protein crystallization to be screened with detergent- containing protein drop.
Catalog No. NC9404328 Supplier Molecular Dimensions Supplier No. MD133
Add to Cart
Add to Cart
Protein Crystallography. MEMSTART + MEMSYS Screen HT-96 (96 x 1.1mL) For transmembrane proteins using vapour diffusion methods, allows the pH range, precipitants and salts used in membrane protein crystallization to be screened with detergent- containing protein drop.