Learn More
Medchemexpress LLC HY-P1276A 1mg Medchemexpress, Men 10376 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1276A1MG
Encompass_Preferred
Medchemexpress, HY-P1276A 1mg Men 10376 (TFA) CAS: Men 10376 TFA is a selective tachykinin NK-2 receptor antagonist, with a K i of 4.4 μM for rat small intestine NK-2 receptor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4180