Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magid™ Men's TouchMaster Cotton Lisle Inspection Gloves - 8 in.
SDP

Catalog No. 19271796
Change view
Click to view available options
Size:
Jumbo
Universal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-271796 Universal
19-271797 Jumbo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-271796 Supplier Magid™ Supplier No. 651
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, two-piece inspection gloves are constructed of cool and breathable cotton. Gloves can be worn alone during delicate applications, or used as a liner to absorb moisture.

  • 100% cotton lisle
  • Breathable, lightweight fabric
  • Can be worn alone or used as glove liners
  • Ideal for general, light-duty inspection
  • Cut & sewn pattern
  • Ambidextrous

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Grade Standard
Hand Ambidextrous
Material Cotton
Product Type Inspection Gloves
Size Universal
For Use With (Application) Light-Duty Inspection
Gender Men's
Series TouchMaster 651
Length (English) 8 in.
Length (Metric) 20.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.