Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Men and Women of Science VHS Video
SCI_ED

Catalog No. S88102
Change view
Click to view available options
Name:
Biology Set of 5 VHS
Chemistry of Burning VHS
Nutrition VHS
Sorting VHS
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Name
S88102 Sorting VHS
S88091 Biology Set of 5 VHS
S88096 Nutrition VHS
S88099 Chemistry of Burning VHS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S88102 Supplier *IMPERIAL INTERNATIONAL Supplier No. II MW 1

missing translation for 'validMainframeMsg'

Includes

Teacher's guide with background information on the scientist, what to watch for questions, discussion topics, and suggestions for follow-up activities. Also includes black line reproducible activity sheets stored in a sturdy vinyl binder.

Behind most major scientific discoveries is a fascinating story.

  • Students in elementary grades and above are treated to a fascinating program that goes beyond coverage of a particular major scientific discovery to a look at the life and work of the scientist behind it
  • These 18 videos offer a unique forum for learning
  • Grades 1-6, remedial for Grades 7-12

Order Info

Sold separately or in topic sets.

Specifications

Grade 7 to 12
Format VHS
Includes Teacher's Guide
Name Sorting VHS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.