Learn More
Aviva Systems Biology Corp Men1 Antibody - middle region (ARP36918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Men1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW045611ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-284-543