Learn More
Cell Signaling Technology Mena (E8H6R) Rabbit Monoclonal Antibody, Endogenous, Rabbit IgG, 100 μL
SDP
Catalog No. 502732879
Encompass
Mena (E8H6R) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total Mena protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human Mena protein. Mena is a member of the Ena/VASP family of proteins, which play a critical role in controlling cell shape and movement by regulating actin dynamics at the growing ends of actin filaments.
  • Validated for Western blotting, immunoprecipitation, and immunofluorescence applications.
  • Recognizes endogenous total protein.
  • Reactivity demonstrated in human, mouse, rat, and monkey models.
  • Supplied in a stabilized glycerol-based formulation.
  • Requires storage at –20°C.
Catalog No. 50-273-2879 Supplier Cell Signaling Technology Supplier No. 57248S
Mena (E8H6R) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total Mena protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human Mena protein. Mena is a member of the Ena/VASP family of proteins, which play a critical role in controlling cell shape and movement by regulating actin dynamics at the growing ends of actin filaments.
  • Validated for Western blotting, immunoprecipitation, and immunofluorescence applications.
  • Recognizes endogenous total protein.
  • Reactivity demonstrated in human, mouse, rat, and monkey models.
  • Supplied in a stabilized glycerol-based formulation.
  • Requires storage at –20°C.