Learn More
Apexbio Technology LLC Menadiol Diacetate 573-20-6 1g
SDP
Catalog No. 5000091814
Encompass_Preferred
Menadiol Diacetate (CAS 573-20-6) is a synthetic derivative of vitamin K investigated for its regulatory effects on cellular signaling pathways and enzyme activities In experimental studies Menadiol Diacetate has been shown to induce S phase arrest in the cell cycle leading to apoptotic cell death This compound is utilized in research focused on cancer and related diseases providing a tool for exploring mechanisms of cell proliferation and apoptosis in vitro
Catalog No. 50-000-91814 Supplier Apexbio Technology LLC Supplier No. C69521000
Menadiol Diacetate (CAS 573-20-6) is a synthetic derivative of vitamin K investigated for its regulatory effects on cellular signaling pathways and enzyme activities In experimental studies Menadiol Diacetate has been shown to induce S phase arrest in the cell cycle leading to apoptotic cell death This compound is utilized in research focused on cancer and related diseases providing a tool for exploring mechanisms of cell proliferation and apoptosis in vitro