Learn More
Selleck Chemical LLC Menadiol Diacetate S4779-1g
SDP
Catalog No. 503110490
Encompass_Preferred
Menadiol Diacetate (Vitamin K4 Acetomenaphthone Kapilin) is a synthetic hydrophilic menadione compound which is clinically used as hemostasis medicine It induces caspase-3-related apoptosis pathway via mitochondrial dysfunction and DNA fragmentation
Catalog No. 50-311-0490 Supplier Selleck Chemical LLC Supplier No. S47791G
Menadiol Diacetate (Vitamin K4 Acetomenaphthone Kapilin) is a synthetic hydrophilic menadione compound which is clinically used as hemostasis medicine It induces caspase-3-related apoptosis pathway via mitochondrial dysfunction and DNA fragmentation