Learn More
Apexbio Technology LLC Menadione, 10mM (in 1mL DMSO). Cas: 58-27-5 MFCD: MFCD00001681. precursor to vitamin K2
SDP
Catalog No. 501151498
Encompass_Preferred
Menadione (vitamin K3), used as a nutritional supplement, is an inhibitor of mitochondrial DNA polymerase γ (pol γ), with an IC 50 value of 6 µM. Pol γ is responsible for all aspects of mitochondrial DNA synthesis, including all replication, recombination of the mitochondrial genome, and repair of mitochondrial DNA damage. Other sizes are available. Please inqury us for quote.
Catalog No. 50-115-1498 Supplier Apexbio Technology LLC Supplier No. B19665.1
 Restricted Item
Add to Cart
Add to Cart
Menadione (vitamin K3), used as a nutritional supplement, is an inhibitor of mitochondrial DNA polymerase γ (pol γ), with an IC 50 value of 6 µM. Pol γ is responsible for all aspects of mitochondrial DNA synthesis, including all replication, recombination of the mitochondrial genome, and repair of mitochondrial DNA damage. Other sizes are available. Please inqury us for quote.