Learn More
Selleck Chemical LLC Menadione 10mM/1mL 58-27-5 Vitamin K3
SDP
Catalog No. 501361795
Encompass_Preferred
Menadione(Vitamin K3), a fat-soluble compound, is an inhibitor of Cdc25 phosphatase and mitochondrial DNA polymerase ? (pol ?), used as a nutritional supplement. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1795 Supplier Selleck Chemical LLC Supplier No. S194910MM/1ML
Add to Cart
Add to Cart
Menadione(Vitamin K3), a fat-soluble compound, is an inhibitor of Cdc25 phosphatase and mitochondrial DNA polymerase ? (pol ?), used as a nutritional supplement. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.