Learn More
Medchemexpress LLC HY-B1897A 1g Medchemexpress, Menadione bisulfite (sodium) CAS:130-37-0 Purity:>98%
SDP
Catalog No. 501933030
Encompass_Preferred
Medchemexpress, HY-B1897A 1g Menadione bisulfite (sodium) CAS:130-37-0 Menadione bisulfite (sodium) is used as an agent to induce acute oxidative stress, and to function as a plant-defense activator against several pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3030 Supplier Medchemexpress LLC Supplier No. HYB1897A1G
Medchemexpress, HY-B1897A 1g Menadione bisulfite (sodium) CAS:130-37-0 Menadione bisulfite (sodium) is used as an agent to induce acute oxidative stress, and to function as a plant-defense activator against several pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.