Learn More
Medchemexpress LLC Menadione bisulfite (sodium) | 130-37-0 | 99.93% | 1 ML
SDP
Catalog No. 5000412288
Encompass_Preferred
Menadione bisulfite (sodium) is utilized as an agent to induce acute oxidative stress and functions as a plant-defense activator against various pathogens. It is intended for research use only.
  • Induces acute oxidative stress.
  • Activates plant defense against several pathogens.
  • Increases ROS and impacts photosynthetic activity in cells at 500 μM concentration.
  • Decreases global and specific antioxidant defenses at 500 μM concentration.
  • Activates defense responses such as PR, ROS, and HR.
  • Enhances local and systemic resistance to phoma stem canker disease.
  • Induces resistance against downy mildew in pearl millet.
  • Acts by priming specific sets of the plant innate-defense repertoire.
  • Upregulates the expression of a gene encoding GRX480 protein.
  • Highly soluble in water and DMSO.
Catalog No. 50-004-12288 Supplier Medchemexpress LLC Supplier No. HYB1897A10MM/1ML
Menadione bisulfite (sodium) is utilized as an agent to induce acute oxidative stress and functions as a plant-defense activator against various pathogens. It is intended for research use only.
  • Induces acute oxidative stress.
  • Activates plant defense against several pathogens.
  • Increases ROS and impacts photosynthetic activity in cells at 500 μM concentration.
  • Decreases global and specific antioxidant defenses at 500 μM concentration.
  • Activates defense responses such as PR, ROS, and HR.
  • Enhances local and systemic resistance to phoma stem canker disease.
  • Induces resistance against downy mildew in pearl millet.
  • Acts by priming specific sets of the plant innate-defense repertoire.
  • Upregulates the expression of a gene encoding GRX480 protein.
  • Highly soluble in water and DMSO.