Learn More
Medchemexpress LLC Menadione bisulfite sodium | 130-37-0 | 99.93% | 500 MG
SDP
Catalog No. 5000410058
Encompass_Preferred
Menadione bisulfite sodium is an agent that induces acute oxidative stress and acts as a plant-defense activator against various pathogens. It impacts photosynthetic activity by increasing reactive oxygen species and decreasing antioxidant defenses. This compound also enhances resistance to diseases like phoma stem canker and downy mildew.
  • Induces acute oxidative stress
  • Activates plant defense mechanisms against pathogens
  • Increases reactive oxygen species
  • Decreases global and specific antioxidant defenses
  • Enhances resistance to phoma stem canker disease
  • Induces resistance against downy mildew in pearl millet
  • Primes specific components of plant innate-defense repertoire
  • Up-regulates gene expression for GRX480 protein
Catalog No. 50-004-10058 Supplier Medchemexpress LLC Supplier No. HYB1897A500MG
Menadione bisulfite sodium is an agent that induces acute oxidative stress and acts as a plant-defense activator against various pathogens. It impacts photosynthetic activity by increasing reactive oxygen species and decreasing antioxidant defenses. This compound also enhances resistance to diseases like phoma stem canker and downy mildew.
  • Induces acute oxidative stress
  • Activates plant defense mechanisms against pathogens
  • Increases reactive oxygen species
  • Decreases global and specific antioxidant defenses
  • Enhances resistance to phoma stem canker disease
  • Induces resistance against downy mildew in pearl millet
  • Primes specific components of plant innate-defense repertoire
  • Up-regulates gene expression for GRX480 protein