Learn More
Selleck Chemical LLC Menadione bisulfite sodium
SDP
Catalog No. 5000004146
Encompass_Preferred
Menadione(Vitamin K3) bisulfite sodium a fat-soluble compound is an inhibitor of Cdc25 phosphatase and mitochondrial DNA polymerase (pol ) used as a nutritional supplement
Catalog No. 50-000-04146 Supplier Selleck Chemical LLC Supplier No. S5953100MG
Menadione(Vitamin K3) bisulfite sodium a fat-soluble compound is an inhibitor of Cdc25 phosphatase and mitochondrial DNA polymerase (pol ) used as a nutritional supplement