Learn More
Medchemexpress LLC Menadione-d3 (Vitamin K3-d3) | 5172-16-7 | 99.1% | 5 MG
SDP
Catalog No. 5000406681
Encompass_Preferred
Menadione-d3 is the deuterium labeled Menadione. Menadione, a synthetic naphthoquinone, can be converted to active vitamin K2 in vivo. This compound is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06681 Supplier Medchemexpress LLC Supplier No. HYB0332S5MG
Menadione-d3 is the deuterium labeled Menadione. Menadione, a synthetic naphthoquinone, can be converted to active vitamin K2 in vivo. This compound is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS