Learn More
Abcam Menadione, Phosphatase inhibitor, 25G
Catalog No. 502706730
Encompass_Preferred
MW 172.18 Da, Purity >99%. Vitamin K produg able to inhibit protein tyrosine phosphatases and induce DNA strand breaks. Able to prevent erlotinib (ab120881) induced cell-cycle arrest. Generates ROS°. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6730 Supplier Abcam Supplier No. AB14127225G
MW 172.18 Da, Purity >99%. Vitamin K produg able to inhibit protein tyrosine phosphatases and induce DNA strand breaks. Able to prevent erlotinib (ab120881) induced cell-cycle arrest. Generates ROS°. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement