Learn More
Medchemexpress LLC Menadione (Standard) | 58-27-5 | 99.91% | 250 MG
SDP
Catalog No. 5000405605
Encompass_Preferred
Menadione (Standard) is the analytical standard of Menadione, intended for research and analytical applications. It is a naphthoquinone that is converted into active vitamin K2 in the body. Menadione is a potential anticancer agent and radiosensitizer.
  • Used for research and analytical applications
  • Converted into active vitamin K2 in the body
  • Potential anticancer agent
  • Potential radiosensitizer
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-004-05605 Supplier Medchemexpress LLC Supplier No. 250MGHYB0332R
Menadione (Standard) is the analytical standard of Menadione, intended for research and analytical applications. It is a naphthoquinone that is converted into active vitamin K2 in the body. Menadione is a potential anticancer agent and radiosensitizer.
  • Used for research and analytical applications
  • Converted into active vitamin K2 in the body
  • Potential anticancer agent
  • Potential radiosensitizer
  • Commonly used in HPLC, GC, and MS experiments