Learn More
Cayman Chemical Menaquinone 4, Purity: ≥98%, CAS: 863-61-6, 100 mg
SDP
Catalog No. 502968427
Encompass_Preferred
A homolog of vitamin K2; formed primarily via conversion of vitamin K1 in vivo; halts the cell cycle at the G1 phase in HepG2, Hep3B, and Huh7 hepatocellular carcinoma cells in a concentration-dependent manner; inhibits IκB, IκBα phosphorylation, and the transcriptional activity of NF-κB
Catalog No. 50-296-8427 Supplier Cayman Chemical Supplier No. 18423100 MG
A homolog of vitamin K2; formed primarily via conversion of vitamin K1 in vivo; halts the cell cycle at the G1 phase in HepG2, Hep3B, and Huh7 hepatocellular carcinoma cells in a concentration-dependent manner; inhibits IκB, IκBα phosphorylation, and the transcriptional activity of NF-κB