Learn More
Cayman Chemical Menaquinone 9, Purity: ≥95%, CAS: 523-39-7, 10 mg
SDP
Catalog No. 502966120
Encompass_Preferred
A vitamin K2 analog; has been found in various bacteria; binds to soluble nitrate reductase; increases plasma prothrombin concentration in vitamin K-deficient rats
Catalog No. 50-296-6120 Supplier Cayman Chemical Supplier No. 1824210 MG
A vitamin K2 analog; has been found in various bacteria; binds to soluble nitrate reductase; increases plasma prothrombin concentration in vitamin K-deficient rats