Learn More
Medchemexpress LLC HY-B2156 100mg Medchemexpress, Menaquinone-4 CAS:863-61-6 Purity:>98%
SDP
Catalog No. 501933087
Encompass_Preferred
Medchemexpress, HY-B2156 100mg Menaquinone-4 CAS:863-61-6 Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also a adjunctive therapy for the pain of osteoporosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3087 Supplier Medchemexpress LLC Supplier No. HYB2156100MG
Medchemexpress, HY-B2156 100mg Menaquinone-4 CAS:863-61-6 Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also a adjunctive therapy for the pain of osteoporosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.