Learn More
Medchemexpress LLC Menaquinone-4-d7 | 1233937-25-1 | 250 UG
SDP
Catalog No. 5000414243
Encompass_Preferred
Menaquinone-4-d7 (Vitamin K2(MK-4)-d7) is the deuterium labeled Menaquinone-4. Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also an adjunctive therapy for the pain of osteoporosis.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Used as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14243 Supplier Medchemexpress LLC Supplier No. 250UGHYB2156S
Menaquinone-4-d7 (Vitamin K2(MK-4)-d7) is the deuterium labeled Menaquinone-4. Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also an adjunctive therapy for the pain of osteoporosis.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Used as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.