Learn More
MOLTOX Molecular Toxicology Inc mEndo Les Agar, 60 mm Plate, 8 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 2140S218
Encompass_Preferred
m Endo Agar, LES is used for enumerating coliforms in water by membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-661