Learn More
Medchemexpress LLC Menin protein, human (His, M327I) | >95.0% | 10UG
SDP
Catalog No. 5000175910
Encompass_Preferred
Recombinant Menin protein (human, M327I) with an N-terminal His tag, expressed in Escherichia coli and provided in microgram quantities for research use. The material has an approximate molecular weight of 68.9 kDa and a reported purity greater than 95% by reducing SDS-PAGE. Recommended storage is at -80°C; avoid repeated freeze-thaw cycles.
Catalog No. 50-001-75910 Supplier Medchemexpress LLC Supplier No. HYP70316310UG
Recombinant Menin protein (human, M327I) with an N-terminal His tag, expressed in Escherichia coli and provided in microgram quantities for research use. The material has an approximate molecular weight of 68.9 kDa and a reported purity greater than 95% by reducing SDS-PAGE. Recommended storage is at -80°C; avoid repeated freeze-thaw cycles.