Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin protein, human (His, M327I) | >95.0% | 50UG
SDP

Catalog No. 5000175872
Encompass_Preferred

Menin protein, human (His, M327I) is a recombinant variant of the human Menin transcriptional regulator expressed in E. coli and supplied as a filtered solution. The protein is provided with an N-terminal His tag, formulated in Tris-HCl/NaCl/glycerol/DTT, and characterized by low endotoxin and defined binding activity to MLL-derived peptides. Store frozen and avoid repeated freeze-thaw cycles for best stability.

  • Recombinant human Menin variant with N-terminal His tag.
  • Expressed in E. coli and supplied as a filtered solution.
  • Formulated in 50 mM Tris-HCl, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Approximately 68.9 kDa molecular weight.
  • Endotoxin level less than 1 EU/μg.
  • Demonstrated binding activity to MLL-derived peptides (Kd ~14.27 nM).
  • Shipped on dry ice and recommended for long-term storage at -80°C.

Catalog No. 50-001-75872 Supplier Medchemexpress LLC Supplier No. HYP70316350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menin protein, human (His, M327I) is a recombinant variant of the human Menin transcriptional regulator expressed in E. coli and supplied as a filtered solution. The protein is provided with an N-terminal His tag, formulated in Tris-HCl/NaCl/glycerol/DTT, and characterized by low endotoxin and defined binding activity to MLL-derived peptides. Store frozen and avoid repeated freeze-thaw cycles for best stability.

  • Recombinant human Menin variant with N-terminal His tag.
  • Expressed in E. coli and supplied as a filtered solution.
  • Formulated in 50 mM Tris-HCl, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Approximately 68.9 kDa molecular weight.
  • Endotoxin level less than 1 EU/μg.
  • Demonstrated binding activity to MLL-derived peptides (Kd ~14.27 nM).
  • Shipped on dry ice and recommended for long-term storage at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.