Learn More
Medchemexpress LLC Menin-MLL inhibitor 19 | 2360487-93-8 | 98.1% | 645.70 | 100 MG
SDP
Catalog No. 5000348201
Encompass_Preferred
Menin-MLL inhibitor 19 is a potent exo-aza spiro inhibitor that targets the menin-mll interaction. This compound is suitable for research into various diseases, including cancer, myelodysplastic syndrome (MDS), and diabetes. It is intended for research use only and not for patient treatment. The product appears as a solid, white to light yellow in color.
  • Potent exo-aza spiro inhibitor of menin-mll interaction
  • Suitable for research on cancer, myelodysplastic syndrome (MDS), and diabetes
  • Appears as a solid
  • White to light yellow color
Catalog No. 50-003-48201 Supplier Medchemexpress LLC Supplier No. HY139076100MG
Menin-MLL inhibitor 19 is a potent exo-aza spiro inhibitor that targets the menin-mll interaction. This compound is suitable for research into various diseases, including cancer, myelodysplastic syndrome (MDS), and diabetes. It is intended for research use only and not for patient treatment. The product appears as a solid, white to light yellow in color.
  • Potent exo-aza spiro inhibitor of menin-mll interaction
  • Suitable for research on cancer, myelodysplastic syndrome (MDS), and diabetes
  • Appears as a solid
  • White to light yellow color