Learn More
Medchemexpress LLC Menin-MLL inhibitor 19 | 2360487-93-8 | 98.1% | 645.70 | 50 MG
SDP
Catalog No. 5000347813
Encompass_Preferred
Menin-MLL inhibitor 19 is a potent exo-aza spiro inhibitor of menin-mll interaction, identified as example A17 from patent WO2019120209A1. This compound is intended for research use only and can be utilized in the research of various diseases, including cancer, myelodysplastic syndrome (MDS), and diabetes.
  • Potent exo-aza spiro inhibitor
  • Used for cancer research
  • Used for myelodysplastic syndrome (MDS) research
  • Used for diabetes research
  • Solid appearance
Catalog No. 50-003-47813 Supplier Medchemexpress LLC Supplier No. HY13907650MG
Menin-MLL inhibitor 19 is a potent exo-aza spiro inhibitor of menin-mll interaction, identified as example A17 from patent WO2019120209A1. This compound is intended for research use only and can be utilized in the research of various diseases, including cancer, myelodysplastic syndrome (MDS), and diabetes.
  • Potent exo-aza spiro inhibitor
  • Used for cancer research
  • Used for myelodysplastic syndrome (MDS) research
  • Used for diabetes research
  • Solid appearance