Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin-MLL inhibitor 19 | 2360487-93-8 | 98.1% | 645.70 | C30H34F3N7O4S | 5 MG
SDP

Catalog No. 5000221332
Encompass_Preferred

Menin-MLL inhibitor 19 is a small-molecule research compound that disrupts the Menin-MLL protein-protein interaction. Supplied as a high-purity solid, it is intended for biochemical and cellular studies of MLL-driven oncogenic and epigenetic pathways.

  • High purity of 98.07% as reported by manufacturer.
  • Molecular weight 645.70 and formula C30H34F3N7O4S.
  • CAS number 2360487-93-8 for unambiguous identification.
  • Appearance: white to light yellow solid, soluble in DMSO.
  • Available in small quantities suitable for screening and dose-response studies.
  • Documentation available: certificate of analysis and safety data sheet.
  • Intended for biochemical and cellular research applications.

Catalog No. 50-002-21332 Supplier Medchemexpress LLC Supplier No. HY1390765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menin-MLL inhibitor 19 is a small-molecule research compound that disrupts the Menin-MLL protein-protein interaction. Supplied as a high-purity solid, it is intended for biochemical and cellular studies of MLL-driven oncogenic and epigenetic pathways.

  • High purity of 98.07% as reported by manufacturer.
  • Molecular weight 645.70 and formula C30H34F3N7O4S.
  • CAS number 2360487-93-8 for unambiguous identification.
  • Appearance: white to light yellow solid, soluble in DMSO.
  • Available in small quantities suitable for screening and dose-response studies.
  • Documentation available: certificate of analysis and safety data sheet.
  • Intended for biochemical and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.