Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin-MLL inhibitor 19 | 2360487-93-8 | 98.1% | C30H34F3N7O4S | 10MG
SDP

Catalog No. 5000261722
Encompass_Preferred

Menin-MLL inhibitor 19 is a small-molecule inhibitor of the menin-MLL protein-protein interaction supplied as a solid for research use; it is used in biochemical and cell-based studies probing menin-mediated oncogenic pathways.

  • Small-molecule menin-MLL interaction inhibitor.
  • Molecular formula C30H34F3N7O4S, molecular weight 645.70 g·mol-1.
  • Appearance: white to light yellow solid.
  • Purity: 98.1% (as supplied).
  • Packaging: 10 mg vial format for research applications.
  • Storage: store sealed away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-61722 Supplier Medchemexpress LLC Supplier No. HY13907610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menin-MLL inhibitor 19 is a small-molecule inhibitor of the menin-MLL protein-protein interaction supplied as a solid for research use; it is used in biochemical and cell-based studies probing menin-mediated oncogenic pathways.

  • Small-molecule menin-MLL interaction inhibitor.
  • Molecular formula C30H34F3N7O4S, molecular weight 645.70 g·mol-1.
  • Appearance: white to light yellow solid.
  • Purity: 98.1% (as supplied).
  • Packaging: 10 mg vial format for research applications.
  • Storage: store sealed away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.