Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin-MLL inhibitor 20 | 2448173-47-3 | 98.8% | 10 MG
SDP

Catalog No. 5000340473
Encompass_Preferred

Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor with demonstrated antitumor activities. This compound is intended for research use only. It has a molecular weight of 612.72 and a chemical formula of C33H40N8O4. It presents as a solid, ranging in color from light yellow to yellow.

  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • For research use only
  • Molecular weight: 612.72
  • Formula: C33H40N8O4
  • Appearance: Solid, light yellow to yellow
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-40473 Supplier Medchemexpress LLC Supplier No. HY12879810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor with demonstrated antitumor activities. This compound is intended for research use only. It has a molecular weight of 612.72 and a chemical formula of C33H40N8O4. It presents as a solid, ranging in color from light yellow to yellow.

  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • For research use only
  • Molecular weight: 612.72
  • Formula: C33H40N8O4
  • Appearance: Solid, light yellow to yellow
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.