Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin-MLL inhibitor 20 | 2448173-47-3 | 98.8% | 100 MG
SDP

Catalog No. 5000339408
Encompass_Preferred

Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor that exhibits antitumor activities. It is referenced in WO2020142557A1, Intermediate 6.

  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • Molecular weight: 612.72
  • Formula: C33H40N8O4
  • Appearance: Light yellow to yellow solid
  • Soluble in DMSO

Catalog No. 50-003-39408 Supplier Medchemexpress LLC Supplier No. HY128798100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor that exhibits antitumor activities. It is referenced in WO2020142557A1, Intermediate 6.

  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • Molecular weight: 612.72
  • Formula: C33H40N8O4
  • Appearance: Light yellow to yellow solid
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.