Learn More
Medchemexpress LLC Menin-MLL inhibitor 20 | 2448173-47-3 | 98.8% | 50 MG
SDP
Catalog No. 5000338567
Encompass_Preferred
Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor with antitumor activities. The Histone-lysine N-methyltransferase 2 (KMT2) family of proteins, including the mixed-lineage leukaemia (MLL) family, methylate lysine 4 on histone H3 tails, which is crucial for modulating chromatin structures and DNA accessibility and plays an important role in gene expression regulation during early development and hematopoiesis. This product is for research use only.
  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • Modulates chromatin structures and DNA accessibility
  • Plays an important role in gene expression regulation
Catalog No. 50-003-38567 Supplier Medchemexpress LLC Supplier No. HY12879850MG
Menin-MLL inhibitor 20 is an irreversible menin-MLL interaction inhibitor with antitumor activities. The Histone-lysine N-methyltransferase 2 (KMT2) family of proteins, including the mixed-lineage leukaemia (MLL) family, methylate lysine 4 on histone H3 tails, which is crucial for modulating chromatin structures and DNA accessibility and plays an important role in gene expression regulation during early development and hematopoiesis. This product is for research use only.
  • Irreversible menin-MLL interaction inhibitor
  • Exhibits antitumor activities
  • Modulates chromatin structures and DNA accessibility
  • Plays an important role in gene expression regulation