Learn More
Medchemexpress LLC HY-15222 5mg Medchemexpress, Menin-MLL inhibitor MI-2 CAS:1271738-62-5 Purity:>98%
SDP
Catalog No. 502081693
Encompass_Preferred
Medchemexpress, HY-15222 5mg Menin-MLL inhibitor MI-2 CAS:1271738-62-5 Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1693 Supplier Medchemexpress LLC Supplier No. HY152225MG
Medchemexpress, HY-15222 5mg Menin-MLL inhibitor MI-2 CAS:1271738-62-5 Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.