Learn More
AdipoGen Mensacarcin
SDP
Supplier:  AdipoGen BVT0028M005
Encompass_Preferred
Chemical. CAS 808750-39-2. Formula C21H24O9. MW 420.4. Isolated from Streptomyces bottropensis. Antibiotic. Antitumor compound. Cytotoxic.
Catalog No. 50-256-2140