Learn More
Cayman Chemical (–)-Menthol, Purity: ≥98%, CAS: 2216-51-5, 5 g
SDP
Catalog No. 502992023
Encompass_Preferred
A monoterpene with diverse biological activities; inhibits AChE (Ki = 1.25 mM for the bovine enzyme); decreases expression of the genes encoding topoisomerase I, IIα, and Iiβ in, and cell viability of, SNU-5 gastric cancer cells at 1.6 mM; reduces acetic acid-induced writhing and increases latency to paw licking in a hot plate test in mice from 3-10 mg/kg
Catalog No. 50-299-2023 Supplier Cayman Chemical Supplier No. 303845 G
A monoterpene with diverse biological activities; inhibits AChE (Ki = 1.25 mM for the bovine enzyme); decreases expression of the genes encoding topoisomerase I, IIα, and Iiβ in, and cell viability of, SNU-5 gastric cancer cells at 1.6 mM; reduces acetic acid-induced writhing and increases latency to paw licking in a hot plate test in mice from 3-10 mg/kg