Learn More
Apexbio Technology LLC Menthol 1490-04-6 200mg
SDP
Catalog No. 5000089222
Encompass_Preferred
Menthol (CAS 1490-04-6) is a widely used small molecule modulator of the TRPM8 ion channel which is implicated in cold sensation By activating TRPM8 menthol induces a cooling response in sensory neurons It also inhibits neuronal calcium channel currents contributing to its analgesic and anti-irritant properties In animal studies menthol has been shown to reduce aversive responses to oral nicotine administration in mice Due to these pharmacological actions menthol serves as a reference standard in biomedical research focused on pain perception sensory modulation and mechanisms of addiction
Catalog No. 50-000-89222 Supplier Apexbio Technology LLC Supplier No. C6774200
Menthol (CAS 1490-04-6) is a widely used small molecule modulator of the TRPM8 ion channel which is implicated in cold sensation By activating TRPM8 menthol induces a cooling response in sensory neurons It also inhibits neuronal calcium channel currents contributing to its analgesic and anti-irritant properties In animal studies menthol has been shown to reduce aversive responses to oral nicotine administration in mice Due to these pharmacological actions menthol serves as a reference standard in biomedical research focused on pain perception sensory modulation and mechanisms of addiction