Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menthol, 99.6%, MP Biomedicals™

Catalog No. ICN15160025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15160025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15160025 Supplier MP Biomedicals, Inc Supplier No. 0215160025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1490-04-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00001484
InChI Key NOOLISFMXDJSKH-UHFFFAOYNA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 1254
ChEBI CHEBI:25187
IUPAC Name 5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)C1CCC(C)CC1O

Specifications

Color Colorless
Density 0.895g/cm3
Assay Percent Range 0.996
Quantity 25 g
Formula Weight 156.3
Physical Form Crystalline Powder
Chemical Name or Material Menthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.