Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-menthol (d-menthol) | 15356-60-2 | MFCD00062983 | 100.0% | 156.27 g/mol | C10H20O | 250 MG
SDP

Catalog No. 5000302734
Encompass_Preferred

(+)-Menthol (D-menthol) is the optically active dextro isomer of menthol used as a research reagent and reference standard in biochemical and antifungal studies. It is offered in solid and solution formats, with high reported purity and defined storage instructions to preserve stability.

  • High purity (99.95%).
  • Molecular formula C10H20O; molecular weight 156.27 g/mol.
  • Available as solid and DMSO solutions in multiple pack sizes.
  • Suitable for biochemical, pharmacological, and antifungal research applications.
  • Powder storage: -20°C (3 years) or 4°C (2 years) for stability.
  • Solvent storage: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-02734 Supplier Medchemexpress LLC Supplier No. 250MGHYW017277
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(+)-Menthol (D-menthol) is the optically active dextro isomer of menthol used as a research reagent and reference standard in biochemical and antifungal studies. It is offered in solid and solution formats, with high reported purity and defined storage instructions to preserve stability.

  • High purity (99.95%).
  • Molecular formula C10H20O; molecular weight 156.27 g/mol.
  • Available as solid and DMSO solutions in multiple pack sizes.
  • Suitable for biochemical, pharmacological, and antifungal research applications.
  • Powder storage: -20°C (3 years) or 4°C (2 years) for stability.
  • Solvent storage: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.