Learn More
Medchemexpress LLC (-)-Menthol (standard) | 2216-51-5 | 99.8% | C₁₀H₂₀O | 100 MG
SDP
Catalog No. 5000434581
Encompass_Preferred
(-)-Menthol is an analytical standard used for research and analytical applications. It is a key component of peppermint oil that binds and activates transient receptor potential melastatin 8 (TRPM8), increasing intracellular calcium. It also exhibits antitumor activity.
  • Used as a reference standard in assays
  • Commonly utilized in qualitative, quantitative, and methodological research
  • Suitable for experiments in HPLC, GC, and MS
  • Appears as a white to off-white solid
  • Found in plants and as an endogenous metabolite
Catalog No. 50-004-34581 Supplier Medchemexpress LLC Supplier No. HY75161R100MG
(-)-Menthol is an analytical standard used for research and analytical applications. It is a key component of peppermint oil that binds and activates transient receptor potential melastatin 8 (TRPM8), increasing intracellular calcium. It also exhibits antitumor activity.
  • Used as a reference standard in assays
  • Commonly utilized in qualitative, quantitative, and methodological research
  • Suitable for experiments in HPLC, GC, and MS
  • Appears as a white to off-white solid
  • Found in plants and as an endogenous metabolite