Learn More
Medchemexpress LLC (-)-Menthol (Standard) | 2216-51-5 | 99.8% | C₁₀H₂₀O | 250 MG
SDP
Catalog No. 5000358034
Encompass_Preferred
(-)-Menthol (Standard) is an analytical standard intended for research and analytical applications. It is a key component of peppermint oil that binds and activates transient receptor potential melastatin 8 (TRPM8), a Ca2+-permeable nonselective cation channel, to increase [Ca2+]i. It also exhibits antitumor activity.
  • Reference standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Key component of peppermint oil
  • Binds and activates TRPM8
  • Exhibits antitumor activity
Catalog No. 50-003-58034 Supplier Medchemexpress LLC Supplier No. 250MGHY75161R
(-)-Menthol (Standard) is an analytical standard intended for research and analytical applications. It is a key component of peppermint oil that binds and activates transient receptor potential melastatin 8 (TRPM8), a Ca2+-permeable nonselective cation channel, to increase [Ca2+]i. It also exhibits antitumor activity.
  • Reference standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Key component of peppermint oil
  • Binds and activates TRPM8
  • Exhibits antitumor activity