Learn More
Medchemexpress LLC (-)-Menthol (Standard) | 2216-51-5 | 99.8% | C₁₀H₂₀O | 50 MG
SDP
Catalog No. 5000434627
Encompass_Preferred
(-)-Menthol (Standard) is the analytical standard of (-)-Menthol, used for research and analytical applications. It is a key component of peppermint oil that binds and activates TRPM8, a Ca2+-permeable nonselective cation channel, increasing [Ca2+]i. This compound also exhibits antitumor activity.
  • Analytical standard for research and analytical applications
  • Key component of peppermint oil
  • Binds and activates TRPM8 to increase [Ca2+]i
  • Exhibits antitumor activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-34627 Supplier Medchemexpress LLC Supplier No. HY75161R50MG
(-)-Menthol (Standard) is the analytical standard of (-)-Menthol, used for research and analytical applications. It is a key component of peppermint oil that binds and activates TRPM8, a Ca2+-permeable nonselective cation channel, increasing [Ca2+]i. This compound also exhibits antitumor activity.
  • Analytical standard for research and analytical applications
  • Key component of peppermint oil
  • Binds and activates TRPM8 to increase [Ca2+]i
  • Exhibits antitumor activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS