Learn More
Medchemexpress LLC Menthone | 10458-14-7 | 500 G
SDP
Catalog No. 5000421390
Encompass_Preferred
Menthone is an orally active monoterpene isolated from plants and Mentha oil. It possesses antibacterial, antitumor, antioxidation, and antivirus properties. As a major volatile component of essential oil, it also exhibits anti-inflammatory properties.
  • Possesses antibacterial, antitumor, antioxidation, and antivirus properties
  • Main volatile component of essential oil
  • Exhibits anti-inflammatory properties in Schistosoma mansoni infection and rheumatoid arthritis
  • Inhibits secretion of proinflammatory cytokine TNF-α
  • Improves allergic inflammatory status of the lungs and airways
Catalog No. 50-004-21390 Supplier Medchemexpress LLC Supplier No. HYN2381500G
Menthone is an orally active monoterpene isolated from plants and Mentha oil. It possesses antibacterial, antitumor, antioxidation, and antivirus properties. As a major volatile component of essential oil, it also exhibits anti-inflammatory properties.
  • Possesses antibacterial, antitumor, antioxidation, and antivirus properties
  • Main volatile component of essential oil
  • Exhibits anti-inflammatory properties in Schistosoma mansoni infection and rheumatoid arthritis
  • Inhibits secretion of proinflammatory cytokine TNF-α
  • Improves allergic inflammatory status of the lungs and airways