Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Menthone | 14073-97-3 | 100 G
SDP

Catalog No. 5000418915
Encompass_Preferred

(-)-Menthone is a monoterpene component found in the essential oil of maturing peppermint. It serves as a precursor to (+)-Neomenthyl-β-d-glucoside and has shown efficacy in affecting the mortality of *Anopheles stephensi* larvae.

  • Purity: 98.0%
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Molecular weight: 154.25
  • Formula: C10H18O
  • Melting/freezing point: -6°C
  • Flash point: 72°C
  • Density: 0.893 g/cm3
  • Classification: Ketones, aldehydes, acids
  • Initial source: Plants other families

Catalog No. 50-004-18915 Supplier Medchemexpress LLC Supplier No. HYN7916100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Menthone is a monoterpene component found in the essential oil of maturing peppermint. It serves as a precursor to (+)-Neomenthyl-β-d-glucoside and has shown efficacy in affecting the mortality of *Anopheles stephensi* larvae.

  • Purity: 98.0%
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Molecular weight: 154.25
  • Formula: C10H18O
  • Melting/freezing point: -6°C
  • Flash point: 72°C
  • Density: 0.893 g/cm3
  • Classification: Ketones, aldehydes, acids
  • Initial source: Plants other families

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.