Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Menthone | 14073-97-3 | 100 MG
SDP

Catalog No. 5000418377
Encompass_Preferred

(-)-Menthone is a monoterpene component found in the essential oil of maturing peppermint. It is metabolized into (+)-Neomenthyl-β-d-glucoside. Studies show that exposure to approximately 45 ppm of (-)-Menthone results in significant mortality in Anopheles stephensi larvae.

  • Purity: 98.0%
  • Molecular weight: 154.25
  • Chemical formula: C10H18O
  • Appearance: Colorless to light yellow liquid
  • Metabolite of peppermint essential oil
  • Exhibits insecticidal activity

Catalog No. 50-004-18377 Supplier Medchemexpress LLC Supplier No. HYN7916100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Menthone is a monoterpene component found in the essential oil of maturing peppermint. It is metabolized into (+)-Neomenthyl-β-d-glucoside. Studies show that exposure to approximately 45 ppm of (-)-Menthone results in significant mortality in Anopheles stephensi larvae.

  • Purity: 98.0%
  • Molecular weight: 154.25
  • Chemical formula: C10H18O
  • Appearance: Colorless to light yellow liquid
  • Metabolite of peppermint essential oil
  • Exhibits insecticidal activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.