Learn More
Medchemexpress LLC HY-N2381 500mg Medchemexpress, Menthone CAS:10458-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2381500MG
Encompass_Preferred
Medchemexpress, HY-N2381 500mg Menthone CAS:10458-14-7 Menthone, a monoterpene extracted from plants and Mentha oil with strong antioxidant properties. Menthone is a main volatile component of the essential oil, and has anti-Inflammatory properties in Schistosoma mansoni Infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2892