Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Menthoxyacetyl Chloride 98.0+%, TCI America™
SDP

Catalog No. M057110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M057110G Supplier TCI America Supplier No. M057110G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15356-62-4
Molecular Formula C12H21ClO2
Molecular Weight (g/mol) 232.75
MDL Number MFCD00044947
InChI Key VNMCKLVHDJADEB-UHFFFAOYNA-N
PubChem CID 11009846
IUPAC Name 2-{[5-methyl-2-(propan-2-yl)cyclohexyl]oxy}acetyl chloride
SMILES CC(C)C1CCC(C)CC1OCC(Cl)=O

Specifications

Color Yellow
Boiling Point 136°C
Quantity 10 g
UN Number 3265
Formula Weight 232.75
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (-)-Menthoxyacetyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.