Learn More
Description
- Menthyl acetate (ma), a monoterpene is one of the main constituent of the peppermint essential oil obtained from the leaves of mentha piperita
- It has been reported to show ambulation-promoting effect in mice
- Its fumigant and repellent activity has been evaluated on first-instar nymphs of the bloodsucking bug rhodnius prolixus
- The decomposition of ma in an atmospheric flow reactor and a low-pressure flat flame has been investigated by molecular-beam mass spectrometry (mbms).menthyl acetate may be used in the preparation of 3-amino-2-hydroxyalkanoates via lithium amide-supported mannich-type reaction of its lithium enolate with pmp-arylaldimines (pmp=p-methoxyphenyl) followed by in situ oxidation with oxaziridine
Specifications
Specifications
| CAS | 89-48-5 |
| Density | 0.922 g/mL (at 25°C (literature)) |
| Boiling Point | 228°C to 229°C (lit.) |
| Molecular Formula | C12H22O2 |
| Refractive Index | n20/D 1.447 (literature) |
| Linear Formula | C12H22O2 |
| MDL Number | MFCD00062975 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 198.3 |
| Percent Purity | 97% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.