Learn More
Medchemexpress LLC Menthyl isovalerate | 16409-46-4 | MFCD00045488 | 98.0% | 240.38 g/mol | C15H28O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW683864500MG
Encompass_Preferred
Menthyl isovalerate is an ester of menthol and isovaleric acid used as a food additive for flavor and fragrance. It is supplied as a colorless to light yellow liquid with a typical purity of 98.0% and a molecular weight of 240.38 g/mol, suitable for formulation and analytical use.
  • Used as a flavor and fragrance additive in food applications.
  • High purity: 98.0%.
  • Colorless to light yellow liquid with a fruity, cooling odor.
  • Molecular formula C15H28O2 and molecular weight 240.38 g/mol.
  • Identified by CAS number 16409-46-4.
  • Packaged in small quantities for research and formulation use.
Catalog No. 50-002-83949